Dichlor(methyl)silan ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97+%
CAS No.: 75-54-7
Molecular Formula: CH4Cl2Si
Formula Weight: 115.03
Flash Point: -32°(-26°F)
Physical Form: Liquid
Density: 1.105
Refractive Index: 1.3980
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Melting Point: -93°
Boiling Point: 41-42°
Beilstein Reference: 1071194
UN No.: 1242
MDL No.: MFCD00000494
Formel:
CH₄Cl₂Si Siedepunkt: 41…42 °C Schmelzpunkt: –93 °C Dichte: 1,105 Flammpunkt: –32 °C (–26 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000494 CAS-Nummer: 75-54-7 EINECS: 200-877-1 UN: 1242 ADR: 4.3,I REACH: 01-2119436669-21 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...