Tetraethylorthosilikat (TEOS) ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
ortho-Kieselsäuretetraethylester
14082.18
14082.36
14082.A4
14082.57
14082.18EA
21.7
EUR
14082.18
14082.36
14082.A4
14082.57
Tetraethylorthosilikat (TEOS) ≥99%
Tetraethylorthosilikat (TEOS)
Purity: 99+%
CAS No.: 78-10-4
Molecular Formula: C8H20O4Si
Formula Weight: 208.33
Flash Point: 46°(116°F)
Physical Form: Liquid
Density: 0.934
Refractive Index: 1.3820
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pleasant
Melting Point: -85°
Boiling Point: 168°
Beilstein Reference: 1422225
Merck Reference: 14,3851
UN No.: 1292
MDL No.: MFCD00009062
As a binding agent and as a polymerization catalyst
Formel:
Si(OC₂H₅)₄ Molecular Weight: 208.33 g/mol Siedepunkt: 168 °C Schmelzpunkt: –85 °C Dichte: 0.934 Flammpunkt: 46 °C (116 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009062 CAS-Nummer: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3 III Merck Index: 13,03882 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...