Reinigungswagen, 2- und 3-Eimer-System, SlimLine™
Lieferant: MICRONOVA
C-22S C-33S
141-0104EA 5290 EUR
141-0104 141-0105
Reinigungswagen, 2- und 3-Eimer-System, SlimLine™
Mopps
Durable cart systems feature a stainless steel frame with push bar and three grip-it holders for easy storage and interchange of handles and additional mop heads. Systems come standard with colour coded, 22,7 L (6 gal.) polypropylene buckets.
  • Crafted from 304 electropolished stainless steel for durability and long life
  • Angled push bar for easy handling in the cleanroom
  • Colour coded, 22 L (6 gallon) polypropylene, or 30 L (8 gallon) stainlesss steel buckets included
  • Optional wringer bar allows wringers to be placed above the rinse bucket
  • Prevenz™ antimicrobial wheels
  • Foot activated brake, entire unit is autoclavable
  • Buckets are best autoclaved in the inverted position
Micronova SlimLine™ stainless steel cart systems, with either polypropylene plastic or stainless steel buckets, accommodate a range of mop wringers. An optional wringer bar, designed for use with the W-6 down press wringer, allows placement above the rinse or empty third bucket for simplified wringing.
Model C-227 includes two 30 litre (8 gal.) buckets. MICRC-22S and MICRC-33S carts include the 6 gallon plastic buckets. The C-227 includes two 8 gallon buckets, made from moulded stainless steel with no seams or ridges to harbour bacteria.
Zubehörinformationen: Buckets are included with carts.
Bestellinformation: Replacement buckets and replacement wheels (RP-05) are also available. All other components must be ordered separately.