Flexible dusting wand, HYGEN™ Quick-Connect
Lieferant: Rubbermaid Commercial Products
FGQ85000BK00 FGQ85100GR00
141-0131EA 38.1 EUR
141-0131 141-0132
Flexible dusting wand, HYGEN™ Quick-Connect
Mopps
Removes dust and allergens from crevices and hard-to-reach areas. Quick connect handle converts dusting wand to an extended reach cleaning tool.
  • Can be laundered
  • Quick connect handle easily converts wand to extended reach cleaning tool
Bitte beachten For dry dusting only—do not use wet.