Reinraum-Staubsauger, MV-1CR (CG), Tiger-Vac®
Lieferant: TIGER-VAC
110739B
141-0955EA 2660 EUR
141-0955
Reinraum-Staubsauger, MV-1CR (CG), Tiger-Vac®
Staubsauger
Designed for general clean up in contamination controlled environments. Cleanroom compatible, can be used in the pharmaceutical industry.
Contruction: Stainless steel AISI 304.
  • Four-stage filtration system includes a ULPA filter (U15); double cloth filter, dome filter and disposable filter bag
  • Static-free
  • Special filter lined with stainless steel is available for the recovery of broken wafers and sharp objects
  • Single phase through-flow motor
  • Recovery capacity of 5,7 litres in disposable filter bag
  • Equipped with a golf cart for easy maneuverability