Cleanroom wipes, ProChlor SATWIPES®
Lieferant: CONTEC
FSWPP02PC
141-1381EA 999 EUR
141-1381
Cleanroom wipes, ProChlor SATWIPES®
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
ProChlor is a sporicide achieving a 100% kill against spores in under one minute. It is validated for sporicidal transfer disinfection in practice-mimicking conditions, achieving a log 5 kill on syringe paper samples and complete kill on plastic syringe samples in one minute.
  • Effective against bacteria, fungi, moulds, yeasts, viruses and spores
  • Contains no quaternary ammonium or surfactant makes it have very low residue, saving time on residue removal
  • Linear tear bags are easy to open even when wearing gloves
ProChlor can be added directly to the dry roll, 5 mins before use, and use the pre-saturated wipes within 8 hours.
ProChlor is 0,2 μm aseptically filtered, and filled in a Grade A environment, into pre-irradiated components. The dry wipe canisters are irradiated at no less than 25 kGy.
Lieferumfang: Each double bag pack contains a canister of 120×34 g/m² melt blown polypropylene wipes and 700 ml ProChlor, in a 1 l bottle.
Verpackung: Double bagged for ease of use in pharmaceutical cleanrooms.
Bitte beachten Use biocides safely. Always read the label and product information before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904
*Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904