Disinfectant, ClearKlens Tego 2000 SC, VH25
Lieferant: Diversey
100868202
141-1447EA 441 EUR
141-1447
Disinfectant, ClearKlens Tego 2000 SC, VH25
Reinigungsmittel Desinfektionsmittel
ClearKlens Tego 2000 SC is a concentrated disinfectant for the application in the pharmaceutical and medical industries.
  • Sterile, amphoteric disinfectant
  • Detex label for proof of irradiation
  • Triple packed to facilitate transport to the clean room
  • No discolouration, low toxicity and odourless
  • Broad spectrum of efficacy
  • Effective in hard and soft water
ClearKlen's Tego 2000 SC is 0,2 μm filtered and triple packaged, validated gamma irradiation process. It is suitable for spray and nebulisation applications.
Bitte beachten Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.