BucketBinders™ Multi Bucket Systems
Lieferant: MICRONOVA
BB-DBL-DBYE BB-TR-DBREYE BB-DBL-DGRE
141-1470EA 866 EUR
141-1470 141-1471 MICRBB-DBL-DGRE
BucketBinders™ Multi Bucket Systems
Mopps
BucketBinders™ bucket system is a cost effective, ergonomic and simple that replaces the need for bulky carts.
  • Easy clip action allows attachment before or after bucket is filled with disinfectant
  • Polyketone hooks firmly secure two or three rectangular buckets together
  • Stainless steel base plate attaches at caster for support
  • Sturdy enough to secure two or three buckets and easy to maneuver with up to 15 litres of liquid in each bucket
  • Lightweight; less than 1 kg per pair
  • Allows scale up or down as required
  • Autoclavable
The system offers easy movement, cleaning, and storage. Works well in both small and large controlled environments and the sterile core.
Will house W-94 flat mop Megawringer™, W-9, W-92 flat wringers or W-9B BoxWringer for string mops. Complements all the Micronova M-Zone FlatMops™, SnapMops™ and SlimLine™ mops.
Zubehörinformationen: B-7 polypropylene buckets with autoclavable castors available in five colours, (blue, green, red, green and grey) are available as replacement buckets.
Verpackung: BucketBinder™ kits include two or three polypropylene buckets with castors (22 L capacity), and one or two sets of BucketBinder bucket connectors.