Calciumoxid, Pulver, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Quicklime, Calcium oxide from marble, caustic, Lime

1410-05EA 359.66 EUR
1410-05 1410-01
Calciumoxid, Pulver, BAKER ANALYZED®, J.T.Baker®
Calcium oxide

Powder. Lot analysis on label.

Formel: CaO
Siedepunkt: 2850 °C (1013 hPa)
Schmelzpunkt: 2580 °C
Dichte: 3,38 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010911
CAS-Nummer: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 1.0 %
Insoluble in CH₃COOH and NH₄OH Precipitate ≤ 1.0 %
Loss on Ignition ≤ 5.0 %
Chloride (Cl) ≤ 0.005 %
Nitrate (NO₃) ≤ 0.05 %
Sulfate (SO₄) ≤ 0.1 %
Heavy Metals (as Pb) ≤ 0.005 %
Iron (Fe) ≤ 0.1 %
Assay, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR