di-Natriummetasilicat Pentahydrat, tech.
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
14103.A1
14103.D9
14103.A1EA
45.4
EUR
14103.A1
14103.D9
di-Natriummetasilicat Pentahydrat, tech.
di-Natriummetasilicat Pentahydrat
CAS No.: 10213-79-3
Molecular Formula: Na2SiO3.5H2O
Formula Weight: 212.14 (122.06anhy)
Physical Form: -18 Mesh Granular
Density: 1.75
Appearance: White
Odour: Odourless
Melting Point: 72°
Merck Reference: 14,8642
UN No.: 3253
MDL No.: MFCD00149176
Formel:
Na₂SiO₃·5H₂O Schmelzpunkt: 72 °C Dichte: 1,75 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149176 CAS-Nummer: 10213-79-3 EINECS: 229-912-9 UN: 3253 ADR: 8,III Merck Index: 14,8642 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...