di-Natriummetasilicat, wasserfrei, körnig, tech. –18 Mesh

Lieferant: Thermo Fisher Scientific
Gefahr

14106.A1 14106.D9 14106.99
14106.A1EA 62.4 EUR
14106.A1 14106.D9 14106.99
di-Natriummetasilicat, wasserfrei, körnig, tech. –18 Mesh
di-Natriummetasilicat

Sodium metasilicate is used as a starting material for zeolites and silica catalysts. It acts as an adhesive and binder, corrosion inhibitor, penetrating sealant, coagulant in waste water treatment, fixative for hand dyeing with reactive dye and detergent auxiliaries.


It is also used in boiler compounds, cleaners, bleaches, aluminum paint strippers and washing carbonated drinking bottles. It finds application in cements, drilling fluids, fireproofing wood, refractories and automobiles. Further, it serves as a surfactant, emulsifier, wetting agent, suspending agent, dispersing agent and anti-corrosive agent in boiler water.


Incompatible with acids, bronze, aluminum, brass, copper, lead, tin, zinc and other alkali sensitive metals.

Formel: Na₂O₃Si
Schmelzpunkt: 1088 °C
Dichte: 2,61
Lagertemperatur: Raumtemperatur
MDL: MFCD00003492
CAS-Nummer: 6834-92-0
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 14,8642

Bestellen

Spezifikation Testergebnisse

Assay (Unspecified), SiO₂ 44.7 - 47.6%
Assay (Unspecified), Na₂O 49.1 - 51.7%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR