Zinkfluorid Tetrahydrat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Zinkdifluorid-Tetrahydrat, Zinkdifluorid Tetrahydrat, Zink(II)fluorid Tetrahydrat
14127.36
14127.36EA
144
EUR
14127.36
Zinkfluorid Tetrahydrat ≥98%
Zinkfluorid Tetrahydrat
Purity: 98%
CAS No.: 13986-18-0
Molecular Formula: ZnF2.4H2O
Formula Weight: 175.43 (103.37anhy)
Physical Form: Crystalline
Odour: Odourless
Merck Reference: 14,10136
UN No.: 3288
MDL No.: MFCD00149892
Formel:
F₂Zn·4H₂O Molecular Weight: 175,45 g/mol Schmelzpunkt: 872 °C Dichte: 2,255 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149892 CAS-Nummer: 13986-18-0 EINECS: 232-001-9 UN: 3288 ADR: 6.1,III Merck Index: 14,10136 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...