Bleititanat ≥99,5%

Lieferant: Thermo Fisher Scientific
Gefahr

14139.22 14139.36 14139.A3
14139.22EA 48.5 EUR
14139.22 14139.36 14139.A3
Bleititanat ≥99,5%
Bleititanat

Purity: 99.5% CAS No.: 12060-00-3 Molecular Formula: PbTiO3 Formula Weight: 303.09 Physical Form: -325 Mesh Powder Density: 7.520 Appearance: Yellow Odour: Odourless UN No.: 3288 MDL No.: MFCD00011159

Formel: O₃PbTi
Schmelzpunkt: 1120 °C
Dichte: 7,520
Lagertemperatur: Raumtemperatur
MDL: MFCD00011159
CAS-Nummer: 12060-00-3
EINECS: 235-038-9
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR