Calciumoxid, Pulver, Laborreagens, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Quicklime, Calcium oxide from marble, caustic, Lime
1414-REA
0
EUR
1414-R
Calciumoxid, Pulver, Laborreagens, J.T.Baker®
Calcium oxide
Formel:
CaO Siedepunkt: 357 °C (1013 hPa) Schmelzpunkt: 169…172 °C Dichte: 1,328 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010911 CAS-Nummer: 1305-78-8 EINECS: 201-064-4 Merck Index: 13,09842 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use |
|
Assay (CaO) | ≥ 90 % |
Insoluble Matter | ≤ 1.0 % |
Loss on Ignition | ≤ 2.0 % |
Carbonate (CO₃) | Passes Test |
Magnesium and Alkali Salts | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...