Tantalpentachlorid ≥99,8% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Tantal(V)chlorid
Unter Argon in Ampullen verpackt
14164.06
14164.14
14164.22
14164.06EA
57.7
EUR
14164.06
14164.14
14164.22
Tantalpentachlorid ≥99,8% (Metall-Basis)
Tantalpentachlorid
Purity: 99.8%
CAS No.: 7721-01-9
Molecular Formula: TaCl5
Formula Weight: 358.21
Physical Form: Powder, Ampouled under argon
Density: 3.68
Appearance: White to pale yellow
Odour: Acidic
Melting Point: 216°
Boiling Point: 242°
Solubility: Decomposes in water. Soluble in absolute alcohol and KOH
Merck Reference: 14,9054
UN No.: 3260
MDL No.: MFCD00011253
In chlorination of organic substances, as an intermediate, and in the production of pure metal
Formel:
Cl₅Ta Siedepunkt: 242 °C Schmelzpunkt: 216 °C Dichte: 3,68 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011253 CAS-Nummer: 7721-01-9 EINECS: 231-755-6 UN: 3260 ADR: 8,III Merck Index: 14,9054 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...