Tantal ≥99,9% (Metall-Basis), Teller, Dicke 6.35 mm (0.25 in)

Lieferant: Thermo Fisher Scientific

14181.FF 14181.FI
14181.FFEA 599 EUR
14181.FF 14181.FI
Tantal ≥99,9% (Metall-Basis), Teller, Dicke 6.35 mm (0.25 in)
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈65.9g/25x25mm Odour: Odourless MDL No.: MFCD00011252

Formel: Ta
Molecular Weight: 180.95 g/mol
Siedepunkt: 5427
Schmelzpunkt: 2996
Dichte: 16.69 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011252
CAS-Nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR