Blei(II,IV)oxid ≥97% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 1314-41-6
Molecular Formula: O4Pb3
Formula Weight: 685.60
Physical Form: Powder
Density: 9.1
Appearance: Bright red to orange
Odour: Odourless
Melting Point: ca 500° dec.
Merck Reference: 14,5425
UN No.: 1479
MDL No.: MFCD00078491
Formel:
Pb₃O₄ Siedepunkt: 2682 °C (1013 hPa) Dichte: 9,1 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078491 CAS-Nummer: 1314-41-6 EINECS: 215-235-6 UN: 1479 ADR: 5.1,II Merck Index: 14,5425 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...