Eisen 99+% Metals basis, wire, Ø 2.0 mm (0.08 in)
Lieferant: Thermo Fisher Scientific
14246.H2
14246.H2EA
784
EUR
14246.H2
Eisen 99+% Metals basis, wire, Ø 2.0 mm (0.08 in)
Iron
Purity: 99+%
CAS No.: 7439-89-6
Physical Form: ≈25g/m
MDL No.: MFCD00010999
Formel:
Fe Molecular Weight: 55.85 g/mol Siedepunkt: 3000 Schmelzpunkt: 1535 Dichte: 7.86 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010999 CAS-Nummer: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...