Wasser, CHROMASOLV™ LC-MS Ultra getestet für UHPLC-MS, Riedel-de Haën™

Lieferant: Honeywell Chemicals

14263-1L 14263-2L
14263-1LEA 80.26 EUR
14263-1L 14263-2L
Wasser, CHROMASOLV™ LC-MS Ultra getestet für UHPLC-MS, Riedel-de Haën™
Wasser

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise

Formel: H₂O
Siedepunkt: 100 °C (1,013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Non-volatile matter Max. 1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Chloride (Cl) Max. 0.01 ppm
Fluoride (F) Max. 0.01 ppm
Nitrate (NO₃) Max. 0.1 ppm
Sulfate (SO₄) Max. 0.1 ppm
UHPLC-MS ESI(+) (Reserpine) Max. 5 ppb
UHPLC-MS ESI(-) (Digoxin) Max. 20 ppb
UHPLC Gradient drift 254 nm Max. 3 mAU
UHPLC Gradient peak 254 nm Max. 0.6 mAU
UHPLC Gradient drift 210 nm Max. 8 mAU
UHPLC Gradient peak 210 nm Max. 2 mAU
Transmittance at 230 nm Min. 99 %
Transmittance at 200 nm Min. 95 %
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
Suitability for the UHPLC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR