Ammoniumformiat, CHROMASOLV™ LC-MS Ultra Eluentzusatz, getestet für UHPLC-MS, Fluka™

Lieferant: Honeywell Chemicals
Achtung

14266-25G
14266-25GEA 247.81 EUR
14266-25G
Ammoniumformiat, CHROMASOLV™ LC-MS Ultra Eluentzusatz, getestet für UHPLC-MS, Fluka™
Ammoniumformiat

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise

Formel: HCO₂NH₄
Schmelzpunkt: 116 °C
Dichte: 1,266 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013103
CAS-Nummer: 540-69-2
EINECS: 208-753-9

Bestellen

Spezifikation Testergebnisse

Assay (calculated on the anhydrous basis) 99.0 - 101.0 %
Water (Karl Fischer) Max. 2 %
Residue on ignition Max. 0.1 %
pH (1 M, 20°C) 5.5 - 7.5
Solution 1 M in H₂O complying
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 0.1 ppm
Barium (Ba) Max. 1 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 5 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 1 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 5 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 1 ppm
Manganese (Mn) Max. 1 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 1 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Suitability for the UHPLC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR