Tantal ≥99,95% (Metall-Basis), dünne Folie, Dicke 0.0127 mm (0.0005 in)
Lieferant: Thermo Fisher Scientific
14266.8S
14266.GD
14266.FY
14266.8SEA
918
EUR
14266.8S
14266.GD
14266.FY
Tantal ≥99,95% (Metall-Basis), dünne Folie, Dicke 0.0127 mm (0.0005 in)
Tantalum
Purity: 99.95%
CAS No.: 7440-25-7
Physical Form: ≈2.1g/100x100mm, certified pinhole free
Odour: Odourless
MDL No.: MFCD00011252
Formel:
Ta Molecular Weight: 180.95 g/mol Siedepunkt: 5427 Schmelzpunkt: 2996 Dichte: 16.69 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011252 CAS-Nummer: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...