Acetonitrile with 0,1% ammonium acetate getestet für UHPLC-MS, Supelco®
Lieferant: Merck
|
Gefahr
|
14274-2L
14274-2LEA
512
EUR
14274-2L
Acetonitrile with 0,1% ammonium acetate getestet für UHPLC-MS, Supelco®
Acetonitrile with 0,1% ammonium acetate
Acetonitrile with ammonium acetate may be used as a mobile phase in liquid chromatography-tandem mass spectrometry (LC-MS/MS) assay for the simultaneous quantitative analysis of synthetic corticosteroids like beclomethasone dipropionate, betamethasone, budesonide, dexamethasone, fludrocortisone, flunisolide, fluorometholone, fluticasone propionate, megestrol acetate, methylprednisolone, prednisolone, prednisone, triamcinolone, and triamcinolone acetonide.
UN:
1648 ADR: 3,II |