Acetonitrile with 0,1% ammonium acetate getestet für UHPLC-MS, Supelco®
Lieferant: Merck
Gefahr
14274-2L
14274-2LEA 512 EUR
14274-2L
Acetonitrile with 0,1% ammonium acetate getestet für UHPLC-MS, Supelco®
Acetonitrile with 0,1% ammonium acetate
Acetonitrile with ammonium acetate may be used as a mobile phase in liquid chromatography-tandem mass spectrometry (LC-MS/MS) assay for the simultaneous quantitative analysis of synthetic corticosteroids like beclomethasone dipropionate, betamethasone, budesonide, dexamethasone, fludrocortisone, flunisolide, fluorometholone, fluticasone propionate, megestrol acetate, methylprednisolone, prednisolone, prednisone, triamcinolone, and triamcinolone acetonide.
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b4a0521b0acf1ca5da766ccc2d70d0d13b96562&vid=7d25b09af109478aea9a2928d6b81c8ff7e88504&prid=web&clid=SAPDAM