Kaliumsulfat ≥99,0% ACS
Lieferant: Thermo Fisher Scientific
14311.36
14311.A3
14311.36EA
41.6
EUR
14311.36
14311.A3
Kaliumsulfat ≥99,0% ACS
Potassium sulphate
Purity: 99.0% min
CAS No.: 7778-80-5
Molecular Formula: K2SO4
Formula Weight: 174.27
Physical Form: Crystalline
Density: 2.662
Melting Point: 1069°
Boiling Point: 1689°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7674
MDL No.: MFCD00011388
Formel:
K₂SO₄ Siedepunkt: 1689 °C Schmelzpunkt: 1069 °C Dichte: 2,662 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011388 CAS-Nummer: 7778-80-5 EINECS: 231-915-5 Merck Index: 14,7674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...