Iod, ultra trocken ≥99,998% (Metall-Basis), Kugeln -10 mesh

Lieferant: Thermo Fisher Scientific
Achtung


Unter Argon in Ampullen verpackt

14312.06 14312.14
14312.06EA 132 EUR
14312.06 14312.14
Iod, ultra trocken ≥99,998% (Metall-Basis), Kugeln -10 mesh
Iod

Iodine spheres are used in OLED materials, chemicals, pharmaceuticals and electronics.


Incompatible with organic materials and alkali metals.


Bitte beachten Store away from reducing agents and metal powders.

Formel: I₂
Siedepunkt: 184,3 °C (1013 hPa)
Schmelzpunkt: 183…186 °C (Subl.)
Dichte: 4,930
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8,III
Merck Index: 13,05036

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Purple
Form Sphere
Total Metal Impurities Max. 0.002%
Particle Size –10 Mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR