Mangan(IV)oxid Mn ≥ 58%, tech., aktiviert
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 1313-13-9
Molecular Formula: MnO2
Formula Weight: 86.94
Physical Form: Powder
Density: 5.026
Appearance: Black
Melting Point: 535° dec.
Solubility: Insoluble in water, nitric acid, sulfuric acid. Soluble in HCl. In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4
Merck Reference: 14,5730
MDL No.: MFCD00003463
As an oxidizing agent, source of metallic manganese, deColourizing glass, painting porcelain, as an analytical reagent
Formel:
MnO₂ Schmelzpunkt: 535 °C (Zersetzung) Dichte: 5,026 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003463 CAS-Nummer: 1313-13-9 EINECS: 215-202-6 Merck Index: 14,5730 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...