Mangan(IV)oxid Mn ≥ 58%, tech., aktiviert

Lieferant: Thermo Fisher Scientific
Achtung

14340.09 14340.22 14340.36
14340.09EA 24.3 EUR
14340.09 14340.22 14340.36
Mangan(IV)oxid Mn ≥ 58%, tech., aktiviert
Mangan(IV)oxid

CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Physical Form: Powder Density: 5.026 Appearance: Black Melting Point: 535° dec. Solubility: Insoluble in water, nitric acid, sulfuric acid. Soluble in HCl. In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4 Merck Reference: 14,5730 MDL No.: MFCD00003463


As an oxidizing agent, source of metallic manganese, deColourizing glass, painting porcelain, as an analytical reagent

Formel: MnO₂
Schmelzpunkt: 535 °C (Zersetzung)
Dichte: 5,026
Lagertemperatur: Raumtemperatur
MDL: MFCD00003463
CAS-Nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,5730

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR