Zinn(II)iodid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
Purity: 99.999%
CAS No.: 10294-70-9
Molecular Formula: SnI2
Formula Weight: 372.50
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.28
Melting Point: 320°
Boiling Point: 720° dec.
Merck Reference: 14,8785
UN No.: 3260
MDL No.: MFCD00049545
Formel:
I₂Sn Siedepunkt: 720 °C (Zers.) Schmelzpunkt: 320 °C Dichte: 5,28 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049545 CAS-Nummer: 10294-70-9 EINECS: 233-667-3 UN: 3260 ADR: 8,III Merck Index: 14,8785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...