di-Ammoniumtellurtetraoxid ≥99,5%

Lieferant: Thermo Fisher Scientific
Achtung

14487.09
14487.09EA 89.1 EUR
14487.09
di-Ammoniumtellurtetraoxid ≥99,5%
di-Ammoniumtellurtetraoxid

Purity: 99.5% CAS No.: 13453-06-0 Molecular Formula: (NH4)2TeO4 Formula Weight: 227.67 Physical Form: -60 Mesh Powder Appearance: White UN No.: 3284 MDL No.: MFCD00015964

Formel: H₈N₂O₄Te
Dichte: 3,02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015964
CAS-Nummer: 13453-06-0
EINECS: 236-622-6
UN: 3284
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR