Natriumnitrat ≥99,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Natronsalpeter

14493.14 14493.30 14493.A1 14493.A7
14493.14EA 32.4 EUR
14493.14 14493.30 14493.A1 14493.A7
Natriumnitrat ≥99,0% ACS
Natriumnitrat

In manufacturing of HNO₃, as a catalyst to manufacture H₂SO₄, in manufacturing of glass, enamel for pottery, sodium nitrite


Purity: 99.0% min CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Physical Form: Crystalline Density: 2.26 Appearance: White Odour: Odourless Melting Point: 308° Solubility: Soluble in water. Slightly soluble in methanol Merck Reference: 14,8647 UN No.: 1498 MDL No.: MFCD00011119


In manufacturing of HNO3, as a catalyst to manufacture H2SO4, in manufacturing of glass, enamel for pottery, sodium nitrite

Formel: NaNO₃
Molecular Weight: 84.99 g/mol
Siedepunkt: 380
Schmelzpunkt: 308
Dichte: 2.26
Lagertemperatur: Raumtemperatur
MDL: MFCD00011119
CAS-Nummer: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Merck Index: 14,8647

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR