Kupfer(II)hexafluoracetylacetonat Hydrat

Lieferant: Thermo Scientific
Achtung

Synonyme: Kupfer(II)Hexafluoro-2,4-pentanedionat Hydrat

14505.03
14505.03EA 41.8 EUR
14505.03
Kupfer(II)hexafluoracetylacetonat Hydrat
Kupfer(II)hexafluoracetylacetonat Hydrat

CAS No.: 155640-85-0 Molecular Formula: Cu(CF3COCHCOCF3)2.xH2O Formula Weight: 477.64(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: ca 100° Boiling Point: 220° dec. Solubility: Soluble in methanol, acetone, toluene MDL No.: MFCD00151019

Formel: C₁₀H₂CuF₁₂O₄·xH₂O
Molecular Weight: 477,64 g/mol (anhy)
Siedepunkt: 220 °C (Zersetzung)
Schmelzpunkt: ∼ 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151019
CAS-Nummer: 155640-85-0
EINECS: 238-850-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR