Ammoniumsulfamat ≥98% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Amidoschwefelsäure Ammoniumsalz, Sulfamidsäure Ammoniumsalz, Ammoniumamidosulfonat, Aminosulfosäure Ammoniumsalz, Ammoniumsulfamidat, Amidosulfonsäure Ammoniumsalz
Purity: 98% min
CAS No.: 7773-06-0
Molecular Formula: H6N2O3S
Formula Weight: 114.12
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odour: Weak
pH: 5.2
Melting Point: 131-135°
Boiling Point: 160° dec.
Solubility: Very soluble in water, liquid ammonia. Slightly soluble in ethanol, glycerol, glycol
Merck Reference: 14,554
MDL No.: MFCD00011429
In electroplating solutions, making nitrous oxide gas, for manufacture of flameproofing textiles and paper products
Formel:
H₆N₂O₃S Siedepunkt: 160 °C (Zersetzung) Schmelzpunkt: 131…135 °C Dichte: >1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011429 CAS-Nummer: 7773-06-0 EINECS: 231-871-7 Merck Index: 14,554 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...