Eisen(II)bromid, ultra trocken ≥99,995% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Eisendibromid
Unter Argon in Ampullen verpackt
14541.03
14541.03EA
110
EUR
14541.03
Eisen(II)bromid, ultra trocken ≥99,995% (Metall-Basis)
Eisen(II)bromid
Purity: 99.995%
CAS No.: 7789-46-0
Molecular Formula: FeBr2
Formula Weight: 215.67
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.636
Melting Point: 684°
Merck Reference: 14,4042
UN No.: 3260
MDL No.: MFCD00016081
Formel:
Br₂Fe Molecular Weight: 215,66 g/mol Schmelzpunkt: 684 °C Dichte: 4,636 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016081 CAS-Nummer: 7789-46-0 EINECS: 232-168-8 UN: 3260 ADR: 8,III Merck Index: 14,4042 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...