Organic reference standard, 4-Methyl-1H-benzotriazole, analytical standard, Supelco®
Lieferant: Merck
14593-50MG
14593-50MGEA 62.5 EUR
14593-50MG
Organic reference standard, 4-Methyl-1H-benzotriazole, analytical standard, Supelco®
Standards Organische Standards
4-Methyl-1H-benzotriazole is an additive in formulated aircraft deicing and anti-icing fluids (ADAFs) mixture. It was used as analytical standard for a study demonstrated to determine benzotriazoles and benzothiazoles in human urine samples using LC- ESI(+)MS/MS.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b754f0c85d8c4a13864a503e9262de5aeaec86a7&vid=bcebb413842470369008984389bcc6abce97ca40&prid=web&clid=SAPDAM