Terbiumtriacetat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
Synonyme:
Terbium(III) acetate hydrate
14600.09
14600.18
14600.09EA
143
EUR
14600.09
14600.18
Terbiumtriacetat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Terbiumtriacetat Hydrat
Purity: 99.9%
CAS No.: 100587-92-6
Molecular Formula: C6H9O6Tb.xH2O
Formula Weight: 336.08(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00149233
Formel:
C₆H₉O₆Tb·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149233 CAS-Nummer: 100587-92-6 EINECS: 247-065-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...