Samarium(III) acetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
Purity: 99.9%
CAS No.: 17829-86-6
Molecular Formula: Sm(OOCCH3)3.xH2O
Formula Weight: 327.48(anhy)
Physical Form: Crystalline Aggregates
Density: 1.94
Sensitivity: Hygroscopic
Odour: Odourless
Boiling Point: 300°
Solubility: Soluble in water
MDL No.: MFCD00150124
Formel:
Sm(OOCCH₃)₃·xH₂O Siedepunkt: 300 °C Dichte: 1,94 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150124 CAS-Nummer: 17829-86-6 EINECS: 233-950-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...