Praseodymium(III) acetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
14611.14
14611.22
14611.14EA
77.9
EUR
14611.14
14611.22
Praseodymium(III) acetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymium(III) acetate hydrate
Purity: 99.9%
CAS No.: 6192-12-7
Molecular Formula: Pr(OOCCH3)3.xH2O
Formula Weight: 318.03(anhy)
Physical Form: Crystalline Aggregates
MDL No.: MFCD00150121
Formel:
Pr(OOCCH₃)₃·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150121 CAS-Nummer: 6192-12-7 EINECS: 228-242-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...