Lanthan(III)isopropoxid (40% La)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 19446-52-7
Molecular Formula: C9H21LaO3
Formula Weight: 316.18
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odour: Alcohol-like
Melting Point: 120-128°
Boiling Point: 170°/0.04mm subl.
UN No.: 2925
MDL No.: MFCD00015305
Formel:
C₉H₂₁LaO₃ Siedepunkt: 170 °C (0,04 mmHg) (Subl.) Schmelzpunkt: 120…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015305 CAS-Nummer: 19446-52-7 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...