Zirkonium ≥99,2% Metall-Basis ausgeschlossen Hf, Drahtform (max. 4,5% Hf), Ø 1.14 mm (0.045 in)
Lieferant: Thermo Fisher Scientific
14627.G2
14627.G6
14627.H2
14627.G2EA
62.3
EUR
14627.G2
14627.G6
14627.H2
Zirkonium ≥99,2% Metall-Basis ausgeschlossen Hf, Drahtform (max. 4,5% Hf), Ø 1.14 mm (0.045 in)
Zirkonium
Purity: 99.2%
CAS No.: 7440-67-7
Physical Form: ≈6.64g/m
Odour: Odourless
MDL No.: MFCD00011303
Formel:
Zr Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011303 CAS-Nummer: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...