Titan(IV)oxid rutil ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.99%
CAS No.: 1317-80-2
Molecular Formula: TiO2
Formula Weight: 79.90
Physical Form: Powder
Density: 4.23
Appearance: White
Odour: Odourless
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formel:
O₂Ti Siedepunkt: 2500…3000 °C Schmelzpunkt: 1830…1850 °C Dichte: 4,23 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011269 CAS-Nummer: 1317-80-2 EINECS: 215-282-2 Merck Index: 14,9472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...