Antimon ≥99,999% (Metall-Basis), Pulver -200 mesh

Lieferant: Thermo Fisher Scientific
Gefahr

14640.03 14640.06 14640.14
14640.03EA 40.6 EUR
14640.03 14640.06 14640.14
Antimon ≥99,999% (Metall-Basis), Pulver -200 mesh
Antimony

Purity: 99.999% CAS No.: 7440-36-0 Odour: Odourless UN No.: 2871 MDL No.: MFCD00134030

Formel: Sb
Lagertemperatur: Raumtemperatur
MDL: MFCD00134030
CAS-Nummer: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR