Kupfer(I)chlorid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.999%
CAS No.: 7758-89-6
Molecular Formula: CuCl
Formula Weight: 98.99
Physical Form: Powder
Density: 4.14
Refractive Index: 1.93
Sensitivity: Air and Moisture Sensitive
Appearance: White
Melting Point: 430°
Boiling Point: 1490°
Merck Reference: 14,2660
UN No.: 2802
MDL No.: MFCD00010971
Formel:
ClCu Siedepunkt: 1490 °C Schmelzpunkt: 430 °C Dichte: 4,14 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010971 CAS-Nummer: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8,III Merck Index: 14,2660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...