Supelco® Organic Reference Standard, Potassium Phthalate Monobasic
Lieferant: Merck
14659-1G
14659-1GEA
131
EUR
14659-1G
Supelco® Organic Reference Standard, Potassium Phthalate Monobasic
Standards
Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.