Calciumdisilicid Ca ∼30% may contain up to 5% Fe, tech.
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Calcium silicide
14676.22
14676.22EA
61.4
EUR
14676.22
Calciumdisilicid Ca ∼30% may contain up to 5% Fe, tech.
Calcium disilicide
CAS No.: 12013-56-8
Molecular Formula: CaSi2
Formula Weight: 96.25
Physical Form: Powder
Density: 2.5
Sensitivity: Moisture Sensitive
Appearance: Grey
Melting Point: ca 1000°
Solubility: Insoluble in cold water, decomposes in hot water
UN No.: 1405
MDL No.: MFCD00015977
Formel:
CaSi₂ Molecular Weight: 96,25 g/mol Schmelzpunkt: ∼ 1000 °C Dichte: 2,5 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015977 CAS-Nummer: 12013-56-8 EINECS: 234-588-7 UN: 1405 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...