Germanium(IV)ethoxid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 14165-55-0
Molecular Formula: C8H20GeO4
Formula Weight: 252.84
Flash Point: 49°(121°F)
Physical Form: Liquid
Density: 1.14
Refractive Index: 1.4070
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Alcohol-like
Melting Point: -72°
Boiling Point: 185.5°
Solubility: Soluble in anhydrous alcohol and benzene. Hydrolyzes in water
UN No.: 1993
MDL No.: MFCD00015121
Formel:
C₈H₂₀GeO₄ Siedepunkt: 185,5 °C Schmelzpunkt: –72 °C Dichte: 1,14 Flammpunkt: 49 °C (121 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015121 CAS-Nummer: 14165-55-0 EINECS: 238-007-8 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...