Niob(V)ethanolat ≥99,999% (Metall-Basis) (max. 0,05% Ta)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.999%
CAS No.: 3236-82-6
Molecular Formula: C10H25NbO5
Formula Weight: 318.21
Flash Point: 36°(97°F)
Physical Form: Liquid
Density: 1.258
Refractive Index: 1.5160
Sensitivity: Moisture Sensitive
Melting Point: 6°
Boiling Point: 140-142°/0.1mm
Solubility: Soluble in organic solvents. Decomposes in water
Beilstein Reference: 3678883
UN No.: 2924
MDL No.: MFCD00015122
Preparation of niobium oxide and mixed metal oxides
Formel:
C₁₀H₂₅NbO₅ Siedepunkt: 140…142 °C (0,1 mmHg) Schmelzpunkt: 6 °C Dichte: 1,258 Flammpunkt: 36 °C (97 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015122 CAS-Nummer: 3236-82-6 EINECS: 221-795-2 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...