Bis-(cyclohexanon)-oxaldihydrazon ≥99,0% zur spektrophotometrischen Bestimmung von Cu, Supelco®

Lieferant: Merck

Synonyme: Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone

M-Clarity™ Qualitätsstufe = MQ100

14690-100G 14690-25G
14690-100GEA 259 EUR
14690-100G 14690-25G
Bis-(cyclohexanon)-oxaldihydrazon ≥99,0% zur spektrophotometrischen Bestimmung von Cu, Supelco®
Bis(cyclohexanone)oxalyldihydrazone

Cuprizone (Bis(cyclohexanone)oxaldihydrazone) is a physically unstable compound and is a well-known neurotoxic agent.


In the presence of a copper salt, this ligand catalyzes Ullmann-type C-N bond forming reactions under mild, aqueous conditions.

Formel: C₁₄H₂₂N₄O₂
Molecular Weight: 278.36 g/mol
Schmelzpunkt: 210…214 °C (Lit.)
MDL: MFCD00001659
CAS-Nummer: 370-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76093ba4d3a0061e297c2b80fc59e8bfcce86434&vid=109545c040198f3292ec6c5bdbfbf8be3d247afc&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c93de7340db204f8060f5c8fdbea6b40264616cd&vid=5195e2ff4346397fe059faa8bb465fe98ab4066e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR