Mangan(II)chlorid, ultra trocken ≥99,998% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Manganese dichloride
Unter Argon in Ampullen verpackt
14697.06
14697.14
14697.06EA
104
EUR
14697.06
14697.14
Mangan(II)chlorid, ultra trocken ≥99,998% (Metall-Basis)
Mangan(II)chlorid
Purity: 99.998%
CAS No.: 7773-01-5
Molecular Formula: Cl2Mn
Formula Weight: 125.84
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.977
Appearance: Pink
Odour: Odourless
Melting Point: 650°
Boiling Point: 1190°
Merck Reference: 14,5728
UN No.: 3288
MDL No.: MFCD00011114
Formel:
MnCl₂ Molecular Weight: 125.84 g/mol Siedepunkt: 1190 °C Schmelzpunkt: 650 Dichte: 2.977 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011114 CAS-Nummer: 7773-01-5 EINECS: 231-869-6 UN: 3288 ADR: 6.1 III Merck Index: 14,5728 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...