Kaliumbromid, ultra trocken ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
Purity: 99.9%
CAS No.: 7758-02-3
Molecular Formula: KBr
Formula Weight: 119.01
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.75
Refractive Index: 1.559
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 730°
Boiling Point: 1435°
Merck Reference: 14,7618
MDL No.: MFCD00011358
Formel:
KBr Siedepunkt: 1435 °C Schmelzpunkt: 730 °C Dichte: 2,75 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011358 CAS-Nummer: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...