Silicon tetraacetate ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
14703.18
14703.30
14703.18EA
128
EUR
14703.18
14703.30
Silicon tetraacetate ≥97%
Silicon tetraacetate
CAS No.: 562-90-3
Molecular Formula: Si(OOCCH3)4
Formula Weight: 264.27
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 110°
Boiling Point: 160-170° dec.
UN No.: UN3261
MDL No.: MFCD00026184
Formel:
C₈H₁₂O₈Si Siedepunkt: 160…170 °C (Zersetzung) Schmelzpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026184 CAS-Nummer: 562-90-3 EINECS: 209-239-7 ADR: 8,II Merck Index: 14,8498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...