Natriumhydrogencarbonat 99,7-100,3% ACS
Lieferant: Thermo Fisher Scientific
Achtung
|
Purity: 99.7-100.3%
CAS No.: 144-55-8
Molecular Formula: NaHCO3
Formula Weight: 84.01
Physical Form: Granular
Density: 2.159
Refractive Index: 1.500
Odour: Odourless
pH: 8,1-8,4
Solubility: Soluble in water. Insoluble in alcohol
Beilstein Reference: 4153970
Merck Reference: 14,8583
MDL No.: MFCD00003528
As a source of CO2, in making sodium salts, baking powder, cleaning compounds
Formel:
NaHCO₃ Schmelzpunkt: 270 °C Dichte: 2,159 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003528 CAS-Nummer: 144-55-8 EINECS: 205-633-8 Merck Index: 14,8583 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...